5-Iodo-2,3-dihydrobenzofuran 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-iodobenzo[b]furan

OR0381-1G OR0381-5G
APOSOR0381-1GEA 58 EUR
APOSOR0381-1G APOSOR0381-5G
5-Iodo-2,3-dihydrobenzofuran 95+%
5-Iodo-2,3-dihydrobenzofuran
Formel: C₈H₇IO
Molecular Weight: 246,05 g/mol
Schmelzpunkt: 62…65 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00817051
CAS-Nummer: 132464-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR