1,3-Dimethyl-1H-pyrazol-5-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

OR0417-5G OR0417-25G
APOSOR0417-5GEA 109 EUR
APOSOR0417-5G APOSOR0417-25G
1,3-Dimethyl-1H-pyrazol-5-carbonsäure 98%
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₈N₂O₂
Molecular Weight: 140,1400 g/mol
Schmelzpunkt: 206…213 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00159636
CAS-Nummer: 5744-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR