8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-8-methyl-8-azabicyclo[3.2.1]octane dihydrochloride

OR0430-1G OR0430-250MG
APOSOR0430-1GEA 49.3 EUR
APOSOR0430-1G APOSOR0430-250MG
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
Formel: C₈H₁₈Cl₂N₂
Molecular Weight: 213.15 g/mol
Schmelzpunkt: 290 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD07366481
CAS-Nummer: 646477-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR