2-Ethyl-6-methylphenol

Lieferant: Apollo Scientific
Achtung

OR0472-1G OR0472-5G
APOSOR0472-1GEA 21.9 EUR
APOSOR0472-1G APOSOR0472-5G
2-Ethyl-6-methylphenol
2-Ethyl-6-methylphenol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Siedepunkt: 96…98 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00962292
CAS-Nummer: 1687-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR