2-Ethyl-6-methylphenylisocyanat

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ethyl-6-methylphenylisocyanate

OR0473-10G OR0473-1G OR0473-5G
APOSOR0473-10GEA 206 EUR
APOSOR0473-10G APOSOR0473-1G APOSOR0473-5G
2-Ethyl-6-methylphenylisocyanat
2-Ethyl-6-methylphenylisocyanat

  • May form small amount of sediment.

Formel: C₁₀H₁₁NO
Molecular Weight: 161,2 g/mol
Siedepunkt: 83…84 °C/5mmHg
Dichte: 1,034 g/cm³ (25 °C)
Flammpunkt: 96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013855
CAS-Nummer: 75746-71-3
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR