Ethyl-2-phenylpropionat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-phenylpropanoate

OR0476-1G OR0476-250MG
APOSOR0476-1GEA 55.1 EUR
APOSOR0476-1G APOSOR0476-250MG
Ethyl-2-phenylpropionat
Ethyl-2-phenylpropionat
Formel: C₁₁H₁₄O₂
Molecular Weight: 178.2277 g/mol
Siedepunkt: 122 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00795471
CAS-Nummer: 2510-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR