1-Boc-4-(4-carboxybenzyl)piperazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-(4-Carboxybenzyl)piperazine, 4-(4-Carboxybenzyl)piperazine-1-carboxylic acid tert-butyl ester, 4-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
OR0480-1G
OR0480-5G
APOSOR0480-1GEA
89.9
EUR
APOSOR0480-1G
APOSOR0480-5G
1-Boc-4-(4-carboxybenzyl)piperazine
1-Boc-4-(4-carboxybenzyl)piperazine
Formel:
C₁₇H₂₄N₂O₄ Molecular Weight: 320,39 g/mol Schmelzpunkt: 212…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974012 CAS-Nummer: 479353-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...