2,6-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR0482-250MG OR0482-1G
APOSOR0482-250MGEA 111 EUR
APOSOR0482-250MG APOSOR0482-1G
2,6-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure
2,6-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,198 g/mol
Schmelzpunkt: 165…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00269372
CAS-Nummer: 81438-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR