N-Fmoc-L-leucin
Lieferant: Apollo Scientific
|
Achtung
|
OR0491-500G
APOSOR0491-500GEA
192
EUR
APOSOR0491-500G
N-Fmoc-L-leucin
N-Fmoc-L-leucin
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,4 g/mol Schmelzpunkt: 152…156 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037133 CAS-Nummer: 35661-60-0 EINECS: 252-662-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...