2-Amino-3-hydroxypyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Amino-3-pyridinol
OR0493-100G
OR0493-500G
OR0493-1KG
APOSOR0493-100GEA
24.8
EUR
APOSOR0493-100G
APOSOR0493-500G
APOSOR0493-1KG
2-Amino-3-hydroxypyridin
2-Amino-3-hydroxypyridin
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Schmelzpunkt: 166…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006317 CAS-Nummer: 16867-03-1 EINECS: 240-886-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...