N-Fmoc-L-valin 97%

Lieferant: Apollo Scientific
Achtung

OR0495-500G
APOSOR0495-500GEA 190 EUR
APOSOR0495-500G
N-Fmoc-L-valin 97%
N-Fmoc-L-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,4 g/mol
Schmelzpunkt: 142…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037124
CAS-Nummer: 68858-20-8
EINECS: 272-515-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR