3-(2-Hydroxyethyl)pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Pyridyl)ethanol

OR0520-1G OR0520-5G
APOSOR0520-1GEA 58 EUR
APOSOR0520-1G APOSOR0520-5G
3-(2-Hydroxyethyl)pyridin
3-(2-Hydroxyethyl)pyridine
Formel: C₇H₉NO
Molecular Weight: 123,15 g/mol
Siedepunkt: 176 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00831047
CAS-Nummer: 6293-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR