3-Hydroxy-4-nitrobenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR0535-100G
OR0535-25G
OR0535-500G
APOSOR0535-100GEA
40.6
EUR
APOSOR0535-100G
APOSOR0535-25G
APOSOR0535-500G
3-Hydroxy-4-nitrobenzoesäure
3-Hydroxy-4-nitrobenzoic acid
Formel:
C₇H₅NO₅ Molecular Weight: 183.12 g/mol Schmelzpunkt: 229…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007110 CAS-Nummer: 619-14-7 EINECS: 210-580-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...