4-(1H-Indol-3-yl)butan-2-one

Lieferant: Apollo Scientific

OR0547-1G OR0547-250MG OR0547-2.5G
APOSOR0547-1GEA 65.4 EUR
APOSOR0547-1G APOSOR0547-250MG APOSOR0547-2.5G
4-(1H-Indol-3-yl)butan-2-one
4-(1H-Indol-3-yl)butan-2-one
Formel: C₁₂H₁₃NO
Molecular Weight: 187,24 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047198
CAS-Nummer: 5541-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR