6-Iod-m-toluidin

Lieferant: Apollo Scientific

Synonyme: 3-Amino-4-iodotoluene, 2-Iodo-5-methylaniline

OR0551-100G OR0551-25G
APOSOR0551-100GEA 183 EUR
APOSOR0551-100G APOSOR0551-25G
6-Iod-m-toluidin
6-Iod-m-toluidin
Formel: C₇H₈IN
Molecular Weight: 233,0496 g/mol
Schmelzpunkt: 37…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00833395
CAS-Nummer: 13194-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR