4-Iod-o-toluidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-iodtoluol, 4-Iod-2-methylanilin

OR0552-100G OR0552-25G OR0552-5G
APOSOR0552-100GEA 95.7 EUR
APOSOR0552-100G APOSOR0552-25G APOSOR0552-5G
4-Iod-o-toluidin
4-Iod-o-toluidin
Formel: C₇H₈IN
Molecular Weight: 233,05 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025299
CAS-Nummer: 13194-68-8
EINECS: 236-154-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR