2-[4-(Isobutyl)phenyl]propionyl chloride

Lieferant: Apollo Scientific

Synonyme: 2-(4-Isobutylphenyl)propanoyl chloride

OR0555-1G OR0555-250MG OR0555-500MG
APOSOR0555-1GEA 405 EUR
APOSOR0555-1G APOSOR0555-250MG APOSOR0555-500MG
2-[4-(Isobutyl)phenyl]propionyl chloride
2-[4-(Isobutyl)phenyl]propionyl chloride
Formel: C₁₃H₁₇ClO
Molecular Weight: 224,73 g/mol
Siedepunkt: 120 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01764800
CAS-Nummer: 34715-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR