L(-)-tert-Leucin (H-Tle-OH) 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH
OR0572-100G
OR0572-25G
OR0572-500G
APOSOR0572-100GEA
145
EUR
APOSOR0572-100G
APOSOR0572-25G
APOSOR0572-500G
L(-)-tert-Leucin (H-Tle-OH) 97%
L(-)-tert-Leucin (H-Tle-OH)
Formel:
C₆H₁₃NO₂ Molecular Weight: 131.2 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064218 CAS-Nummer: 20859-02-3 EINECS: 424-750-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...