3-Tolylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methylbenzolboronsäure, Dihydroxy(3-methylphenyl)boran, (3-Methylphenyl)metaborsäure, 3-Methylphenylboronsäure

OR0643-100G OR0643-500G
APOSOR0643-100GEA 84.1 EUR
APOSOR0643-100G APOSOR0643-500G
3-Tolylboronsäure 97%
3-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: 159…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00040198
CAS-Nummer: 17933-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR