5-Nitro-1H-indole-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR0676-1G
OR0676-250MG
APOSOR0676-1GEA
29
EUR
APOSOR0676-1G
APOSOR0676-250MG
5-Nitro-1H-indole-3-carbaldehyde
5-Nitro-1H-indole-3-carbaldehyde
Formel:
C9H6N2O3 Molecular Weight: 190,1600 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01313781 CAS-Nummer: 6625-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...