(4-Chlorphenyl)methylsulfon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chlor-4-(methylsulfonyl)benzol
OR0678-100G
OR0678-10G
OR0678-50G
APOSOR0678-100GEA
98.6
EUR
APOSOR0678-100G
APOSOR0678-10G
APOSOR0678-50G
(4-Chlorphenyl)methylsulfon 98%
(4-Chlorphenyl)methylsulfon
Formel:
C₇H₇ClO₂S Molecular Weight: 190.65 g/mol Schmelzpunkt: 96…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025067 CAS-Nummer: 98-57-7 EINECS: 202-682-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...