Phenylacetamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenylacetamide

OR0700-25G OR0700-100G
APOSOR0700-25GEA 21.9 EUR
APOSOR0700-25G APOSOR0700-100G
Phenylacetamid 98%
Phenylacetamid
Formel: C₈H₉NO
Molecular Weight: 135,17 g/mol
Schmelzpunkt: 156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059193
CAS-Nummer: 103-81-1
EINECS: 203-147-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR