4-Pyridinboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Pyridinylboronsäure, Pyridin-4-boronsäure

OR0727-25G OR0727-5G OR0727-100G
APOSOR0727-25GEA 37.7 EUR
APOSOR0727-25G APOSOR0727-5G APOSOR0727-100G
4-Pyridinboronsäure 97%
4-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122,92 g/mol (anhy)
Schmelzpunkt: >300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01074545
CAS-Nummer: 1692-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR