4-Chinolincarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Chinolin-4-carbonsäure

OR0731-1G OR0731-25G OR0731-5G
APOSOR0731-1GEA 24.8 EUR
APOSOR0731-1G APOSOR0731-25G APOSOR0731-5G
4-Chinolincarbonsäure
4-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173.17 g/mol
Schmelzpunkt: 253…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006782
CAS-Nummer: 486-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR