3-Piperidin-1-ylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Piperidin-1-yl)benzoic acid
OR0764-1G
OR0764-250MG
OR0764-5G
APOSOR0764-1GEA
194
EUR
APOSOR0764-1G
APOSOR0764-250MG
APOSOR0764-5G
3-Piperidin-1-ylbenzoic acid
3-Piperidin-1-ylbenzoic acid
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205.26 g/mol Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659081 CAS-Nummer: 77940-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...