3,5-Dichlorthiophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dichlorphenylmercaptan
OR0770-1G
OR0770-5G
APOSOR0770-1GEA
106
EUR
APOSOR0770-1G
APOSOR0770-5G
3,5-Dichlorthiophenol 97%
3,5-Dichlorthiophenol
Formel:
C₆H₄Cl₂S Molecular Weight: 179,07 g/mol Siedepunkt: 270…272 °C Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041423 CAS-Nummer: 17231-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...