4-Nitrothioanisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Nitro-4-(methylthio)benzol, 4-Nitrophenylmethylsulfid

OR0780-100G OR0780-25G OR0780-500G
APOSOR0780-100GEA 72.5 EUR
APOSOR0780-100G APOSOR0780-25G APOSOR0780-500G
4-Nitrothioanisol 98%
4-Nitrothioanisol
Formel: C₇H₇NO₂S
Molecular Weight: 169,2 g/mol
Schmelzpunkt: 66…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010868
CAS-Nummer: 701-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR