1-(2-Aminoethyl)-4-methylpiperazin 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(4-Methylpiperazin-1-yl)ethanamine, 2-(4-Methyl-piperazin-1-yl)-ethylamine

OR0800-1G OR0800-5G OR0800-10G
APOSOR0800-1GEA 21.9 EUR
APOSOR0800-1G APOSOR0800-5G APOSOR0800-10G
1-(2-Aminoethyl)-4-methylpiperazin 95%
1-(2-Aminoethyl)-4-methylpiperazin
Formel: C₇H₁₇N₃
Molecular Weight: 143.23 g/mol
Siedepunkt: 112 °C/30mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03701701
CAS-Nummer: 934-98-5
EINECS: 213-296-3
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR