1-Boc-4-piperidinessigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid

OR0804-10G OR0804-1G OR0804-5G
APOSOR0804-10GEA 37.7 EUR
APOSOR0804-10G APOSOR0804-1G APOSOR0804-5G
1-Boc-4-piperidinessigsäure
1-Boc-4-piperidinessigsäure
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243.3000 g/mol
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800239
CAS-Nummer: 157688-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR