4-Brom-2-chloranilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR0849-250G
OR0849-25G
OR0849-500G
APOSOR0849-250GEA
132
EUR
APOSOR0849-250G
APOSOR0849-25G
APOSOR0849-500G
4-Brom-2-chloranilin 98%
4-Brom-2-chloranilin
Formel:
C₆H₅BrClN Molecular Weight: 206.47 g/mol Siedepunkt: 264 Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007660 CAS-Nummer: 38762-41-3 EINECS: 254-118-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...