5-Brom-2-chloranilin

Lieferant: Apollo Scientific
Achtung

OR0859-25G OR0859-5G OR0859-100G
APOSOR0859-25GEA 37.7 EUR
APOSOR0859-25G APOSOR0859-5G APOSOR0859-100G
5-Brom-2-chloranilin
5-Brom-2-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
Schmelzpunkt: 145…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06796283
CAS-Nummer: 60811-17-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR