1-Adamantan-1-ylpiperazine

Lieferant: Apollo Scientific
Achtung

OR0860-1G OR0860-5G
APOSOR0860-1GEA 127 EUR
APOSOR0860-1G APOSOR0860-5G
1-Adamantan-1-ylpiperazine
1-Adamantan-1-ylpiperazine
Formel: C₁₄H₂₄N₂
Molecular Weight: 220,36 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01748381
CAS-Nummer: 19984-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR