Phenyl-4-hydroxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
OR0863-25G
OR0863-500G
OR0863-100G
APOSOR0863-25GEA
0
EUR
APOSOR0863-25G
APOSOR0863-500G
APOSOR0863-100G
Phenyl-4-hydroxybenzoat
Phenyl-4-hydroxybenzoat
Formel:
C₁₃H₁₀O₃ Schmelzpunkt: 182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016467 CAS-Nummer: 17696-62-7 EINECS: 241-698-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...