1-(2-chloro-4-nitrophenyl)piperazine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Chloro-4-nitrophenyl)piperazine
OR0871-1G
OR0871-5G
APOSOR0871-1GEA
23.2
EUR
APOSOR0871-1G
APOSOR0871-5G
1-(2-chloro-4-nitrophenyl)piperazine 95%
1-(2-chloro-4-nitrophenyl)piperazine
Formel:
C₁₀H₁₂ClN₃O₂ Molecular Weight: 241.675 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00640776 CAS-Nummer: 114878-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...