4-Methylpiperazine-1-ethanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Hydroxyethyl)-4-methylpiperazine
OR0879-25G
OR0879-100G
OR0879-5G
APOSOR0879-25GEA
62.5
EUR
APOSOR0879-25G
APOSOR0879-100G
APOSOR0879-5G
4-Methylpiperazine-1-ethanol 95%
4-Methylpiperazine-1-ethanol
Formel:
C₇H₁₆N₂O Molecular Weight: 144,22 g/mol Schmelzpunkt: 54…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840691 CAS-Nummer: 5464-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...