1-Methyl-4-piperidin-4-ylpiperazine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Methyl-4-(piperidin-4-yl)piperazine
OR0885-1G
OR0885-250MG
OR0885-5G
APOSOR0885-1GEA
50.9
EUR
APOSOR0885-1G
APOSOR0885-250MG
APOSOR0885-5G
1-Methyl-4-piperidin-4-ylpiperazine
1-Methyl-4-piperidin-4-ylpiperazine
Formel:
C₁₀H₂₁N₃ Molecular Weight: 183,3 g/mol Schmelzpunkt: 53…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03274729 CAS-Nummer: 53617-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...