2-Amino-5-nitro-4-picolin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methyl-5-nitro-2-pyridylamin, 2-Amino-4-methyl-5-nitropyridin
OR0938-5G
OR0938-25G
APOSOR0938-5GEA
21.9
EUR
APOSOR0938-5G
APOSOR0938-25G
2-Amino-5-nitro-4-picolin
2-Amino-5-nitro-4-picolin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153.14 g/mol Schmelzpunkt: 223…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010692 CAS-Nummer: 21901-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...