1-(Piperidine-1-sulphonyl)piperazine

Lieferant: Apollo Scientific
Achtung

OR0954-1G OR0954-250MG
APOSOR0954-1GEA 0 EUR
APOSOR0954-1G APOSOR0954-250MG
1-(Piperidine-1-sulphonyl)piperazine
1-(Piperidine-1-sulphonyl)piperazine
Formel: C₉H₁₉N₃O₂S
Molecular Weight: 233,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973338
CAS-Nummer: 500587-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR