4-(2,4-Dinitrophenyl)butanoic acid

Lieferant: Apollo Scientific

OR0966-10G OR0966-1G OR0966-5G
APOSOR0966-10GEA 1350 EUR
APOSOR0966-10G APOSOR0966-1G APOSOR0966-5G
4-(2,4-Dinitrophenyl)butanoic acid
4-(2,4-Dinitrophenyl)butanoic acid
Formel: C₁₀H₁₀N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD05663835
CAS-Nummer: 52120-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR