8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 98%
Lieferant: Apollo Scientific
Achtung
Synonyme: Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid
OR0969-1G OR0969-5G OR0969-25G
APOSOR0969-1GEA 21.9 EUR
APOSOR0969-1G APOSOR0969-5G APOSOR0969-25G
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 98%
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid
Formel: C₂₁H₂₃NO₆
Molecular Weight: 385,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321015
CAS-Nummer: 166108-71-0