4-Amino-5-(2-thienyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific
Achtung

OR0998-1G OR0998-250MG
APOSOR0998-1GEA 186 EUR
APOSOR0998-1G APOSOR0998-250MG
4-Amino-5-(2-thienyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-thienyl)-4H-1,2,4-triazole-3-thiol
Formel: C₆H₆N₄S₂
Molecular Weight: 198,27 g/mol
Schmelzpunkt: 219…220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00269270
CAS-Nummer: 61019-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR