2-Cyan-4'-methylbiphenyl

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

OR10059-100G OR10059-25G OR10059-5G
APOSOR10059-100GEA 236 EUR
APOSOR10059-100G APOSOR10059-25G APOSOR10059-5G
2-Cyan-4'-methylbiphenyl
2-Cyan-4'-methylbiphenyl
Formel: C₁₄H₁₁N
Molecular Weight: 193,26 g/mol
Schmelzpunkt: 49…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151805
CAS-Nummer: 114772-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR