Benzo[b]thiophene-3-acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (Benzo[b]thiophen-3-yl)acetic acid

OR100752-1G OR100752-5G
APOSOR100752-1GEA 87 EUR
APOSOR100752-1G APOSOR100752-5G
Benzo[b]thiophene-3-acetic acid
Benzo[b]thiophene-3-acetic acid
Formel: C₁₀H₈O₂S
Molecular Weight: 192.24 g/mol
Schmelzpunkt: 108…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051637
CAS-Nummer: 1131-09-5
EINECS: 214-461-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR