4-(4-Bromophenyl)pyrazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR1011-1G
OR1011-25G
OR1011-5G
APOSOR1011-1GEA
105
EUR
APOSOR1011-1G
APOSOR1011-25G
APOSOR1011-5G
4-(4-Bromophenyl)pyrazole 97%
4-(4-Bromophenyl)pyrazole
Formel:
C₉H₇BrN₂ Molecular Weight: 223,08 g/mol Schmelzpunkt: 208…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06800354 CAS-Nummer: 849021-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...