2,5-Thiophendicarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Thiophene-2,5-dicarboxylic acid
OR10148-50G
OR10148-100G
OR10148-500G
APOSOR10148-50GEA
21.9
EUR
APOSOR10148-50G
APOSOR10148-100G
APOSOR10148-500G
2,5-Thiophendicarbonsäure
2,5-Thiophenedicarboxylic acid
Formel:
C₆H₄O₄S Molecular Weight: 172,16 g/mol Schmelzpunkt: 355…360 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016896 CAS-Nummer: 4282-31-9 EINECS: 224-284-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...