2-Amino-3,5-dibrombenzonitril

Lieferant: Apollo Scientific
Achtung

OR1015-100G OR1015-25G
APOSOR1015-100GEA 335 EUR
APOSOR1015-100G APOSOR1015-25G
2-Amino-3,5-dibrombenzonitril
2-Amino-3,5-dibrombenzonitril
Formel: C₇H₄Br₂N₂
Molecular Weight: 275,93 g/mol
Schmelzpunkt: 160…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179571
CAS-Nummer: 68385-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR