5-Chlor-2-thenoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chlor-2-thiophencarbonsäure, 5-Chlorthiophen-2-carbonsäure

OR10153-25G OR10153-5G
APOSOR10153-25GEA 24.8 EUR
APOSOR10153-25G APOSOR10153-5G
5-Chlor-2-thenoesäure
5-Chlor-2-thenoesäure
Formel: C₅H₃ClO₂S
Molecular Weight: 162,6 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041426
CAS-Nummer: 24065-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR