5-Chloro-6-(4-chlorophenyl)-2-methylpyridazin-3(2H)-one 95+%

Lieferant: Apollo Scientific

OR1018-1G OR1018-5G
APOSOR1018-1GEA 246 EUR
APOSOR1018-1G APOSOR1018-5G
5-Chloro-6-(4-chlorophenyl)-2-methylpyridazin-3(2H)-one 95+%
5-Chloro-6-(4-chlorophenyl)-2-methylpyridazin-3(2H)-one
Formel: C₁₁H₈Cl₂N₂O
Schmelzpunkt: 111…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06800421
CAS-Nummer: 849021-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR