Indol-5-carbonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Indol-5-carbonitril
OR10181-25G
OR10181-5G
APOSOR10181-25GEA
37.7
EUR
APOSOR10181-25G
APOSOR10181-5G
Indol-5-carbonitril
Indol-5-carbonitril
Formel:
C₉H₆N₂ Molecular Weight: 142,16 g/mol Schmelzpunkt: 106…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005669 CAS-Nummer: 15861-24-2 EINECS: 239-986-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...