5-Hydroxyindol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol
OR10182-100G
OR10182-25G
OR10182-5G
APOSOR10182-100GEA
435
EUR
APOSOR10182-100G
APOSOR10182-25G
APOSOR10182-5G
5-Hydroxyindol 98%
5-Hydroxyindol
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol Schmelzpunkt: 106…111 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005677 CAS-Nummer: 1953-54-4 EINECS: 217-782-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...